Learn More
Perphenazine Sulfoxide N1,N4-Dioxide (Perphenazine N1,N4,S-Trioxide), Mikromol™
Description
Perphenazine Sulfoxide N1,N4-Dioxide (Perphenazine N1,N4,S-Trioxide)
Specifications
Specifications
| Chemical Name or Material | Perphenazine Sulfoxide N1,N4-Dioxide |
| CAS | 2724727-08-4 |
| Analyte or Component Names | Perphenazine Sulfoxide N1,N4-Dioxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H26 Cl N3 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Perphenazine Sulfoxide N1,N4-Dioxide |
| InChI Formula | InChI=1S/C21H26ClN3O4S/c22-17-6-7-21-19(16-17)23(18-4-1-2-5-20(18)30(21)29)8-3-9-24(27)10-12-25(28,13-11-24)14-15-26/h1-2,4-7,16,26H,3,8-15H2 |
| SMILES | OCC[N+]1([O-])CC[N+]([O-])(CCCN2c3ccccc3S(=O)c4ccc(Cl)cc24)CC1 |
| IUPAC Name | 2-[4-[3-(2-chloro-5-oxophenothiazin-10-yl)propyl]-1,4-dioxidopiperazine-1,4-diium-1-yl]ethanol |
| Show More |