Learn More
Perphenazine N1,N4-Dioxide Hydrochloride, Mikromol™
Description
Perphenazine N1,N4-Dioxide Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Perphenazine N1,N4-Dioxide Hydrochloride |
| CAS | 2724727-09-5 |
| Analyte or Component Names | Perphenazine N1,N4-Dioxide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H26 Cl N3 O3 S . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Perphenazine N1,N4-Dioxide Hydrochloride |
| InChI Formula | InChI=1S/C21H26ClN3O3S.ClH/c22-17-6-7-21-19(16-17)23(18-4-1-2-5-20(18)29-21)8-3-9-24(27)10-12-25(28,13-11-24)14-15-26;/h1-2,4-7,16,26H,3,8-15H2;1H |
| SMILES | Cl.OCC[N+]1([O-])CC[N+]([O-])(CCCN2c3ccccc3Sc4ccc(Cl)cc24)CC1 |
| IUPAC Name | 2-[4-[3-(2-chlorophenothiazin-10-yl)propyl]-1,4-dioxidopiperazine-1,4-diium-1-yl]ethanol;hydrochloride |
| Show More |