Learn More
Perfluorodecanesulfonic acid sodium 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30326309
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326309 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30326309 Supplier Dr. Ehrenstorfer Supplier No. DREA15986581AL100
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Perfluorodecanesulfonic acid sodium salt
CAS 2806-15-7
Analyte or Component Names Perfluorodecanesulfonic acid sodium salt
Shipping Condition Room Temperature
Molecular Formula C10 F21 O3 S . Na
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 1-Decanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heneicosafluoro-, sodium salt (9CI), Sodium perfluorodecanesulfonate, Perfluorodecanesulfonate, sodium salt, PFDS, sodium salt
InChI Formula InChI=1S/C10HF21O3S.Na/c11-1(12,3(15,16)5(19,20)7(23,24)9(27,28)29)2(13,14)4(17,18)6(21,22)8(25,26)10(30,31)35(32,33)34;/h(H,32,33,34);/q;+1/p-1
SMILES [Na+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC Name sodium;1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-henicosafluorodecane-1-sulfonate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 622.13
Quantity 1 mL
Formula Weight 621.9131 g/mol