Learn More
Perfluoro-2-methyl-3-oxahexanoic acid ammonium 50 μg/mL in Methanol/Water, Dr. Ehrenstorfer
Specifications
Specifications
| Quantity | 1 mL |
| Concentration | 50 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol/Water |