Learn More
Pd(NO3)2 for AAS 2g/L 1% HNO3 matrix, Fisher Chemical™
BIOPHARMA
Product Code. 12976173 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12976173 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12976173 Supplier Fisher Chemical Supplier No. MM 005 050
Please to purchase this item. Need a web account? Register with us today!
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Bottle
Physical Form Liquid
Concentration 2 g/L
Type Matrix Modifier
Solvent or Matrix 1% HNO3 matrix
Quantity 50 mL
Chemical Name or Material Pd(NO3)2