missing translation for 'onlineSavingsMsg'
Learn More

Ficoll 400 (White to Off-White Powder/Molecular Biology), Fisher BioReagents™

Product Code. 10468343
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
10g
25g
500g
This item is not returnable. View return policy

Product Code. 10468343

Brand: Fisher BioReagents BP525100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ficoll 400 (White to Off-White Powder/Molecular Biology)

Ficoll 400 is used as a component of Denhardt's Reagent and in gel-loading buffers for DNA gels. It is also used in forming density gradients to separate cells and subcellular components.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26873-85-8
Molecular Formula C15H27ClO12
Molecular Weight (g/mol) 434.819
InChI Key IACFXVUNKCXYJM-AKSHDPDZSA-N
Synonym ficol 400
PubChem CID 88330506
IUPAC Name 2-(chloromethyl)oxirane;(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1C(O1)CCl.C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O

Specifications

Color White
Physical Form Powder/Solid
Quantity 100 g
Formula Weight ≈400,000
Grade Molecular Biology
Packaging Poly Bottle
Specific Rotation Inclusive between 50° (+ or -)
Chemical Name or Material Ficoll™ 400
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.