missing translation for 'onlineSavingsMsg'
Learn More

p-Xylylenebis(triphenylphosphonium bromide), 96%

Product Code. 11433777
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11433777

Brand: Thermo Scientific Alfa Aesar A18241.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 40817-03-6 | C44H38Br2P2 | 788.54 g/mol

p-Xylylenebis(triphenylphosphonium bromide) is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Xylylenebis(triphenylphosphonium bromide) is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40817-03-6
Molecular Formula C44H38Br2P2
Molecular Weight (g/mol) 788.54
MDL Number MFCD00011914
InChI Key ZZQVVCXWFPGKJD-UHFFFAOYSA-L
Synonym 1,4-phenylenebis methylene bis triphenylphosphonium bromide, p-xylylenebis triphenylphosphonium bromide, 1,4-phenylenebis methylene bis triphenylphosphonium dibromide, rarechem fh 1w 0055, phosphonium,1,1'-1,4-phenylenebis methylene bis 1,1,1-triphenyl-, bromide 1:2, triphenyl 4-triphenylphosphaniumyl methyl phenyl methyl phosphanium dibromide, triphenyl-4-triphenylphosphaniumylmethyl phenyl methyl phosphanium dibromide
PubChem CID 2723797
SMILES [Br-].[Br-].C(C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 3584592
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
IUPAC Name triphenyl({4-[(triphenylphosphaniumyl)methyl]phenyl}methyl)phosphanium dibromide
Formula Weight 788.55
Percent Purity 96%
Chemical Name or Material p-Xylylenebis(triphenylphosphonium bromide)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.