accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 40817-03-6

BrBrPP

CAS RN 40817-03-6

IUPAC Name: triphenyl({4-[(triphenylphosphaniumyl)methyl]phenyl}methyl)phosphanium dibromide
Synonyms: triphenyl({4-[(triphenylphosphaniumyl)methyl]phenyl}methyl)phosphanium dibromide
Molecular Weight (g/mol): 788.54
Molecular Formula: C44H38Br2P2
InChi Key: ZZQVVCXWFPGKJD-UHFFFAOYSA-L
SMILES: [Br-].[Br-].C(C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results