missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Xylylenebis(triphenylphosphonium bromide), 96%
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
p-Xylylenebis(triphenylphosphonium bromide) is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Xylylenebis(triphenylphosphonium bromide) is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
p-Xylylenebis(triphenylphosphonium bromide) is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 25 g |
| Beilstein | 3584592 |
| Sensitivity | Hygroscopic |
| Solubility Information | Slightly soluble in water. |
| IUPAC Name | triphenyl-[[4-(triphenylphosphaniumylmethyl)phenyl]methyl]phosphanium;dibromide |
| Formula Weight | 788.55 |
| Percent Purity | 96% |
| Chemical Name or Material | p-Xylylenebis(triphenylphosphonium bromide) |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction