Learn More
Oxybuprocaine N-Oxide Dihydrochloride, Mikromol™
Description
Oxybuprocaine N-Oxide Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | Oxybuprocaine N-Oxide Dihydrochloride |
| CAS | 2727060-44-6 |
| Analyte or Component Names | Oxybuprocaine N-Oxide Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H28 N2 O4 . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Oxybuprocaine N-Oxide Dihydrochloride |
| InChI Formula | InChI=1S/C17H28N2O4.2ClH/c1-4-7-11-22-16-13-14(8-9-15(16)18)17(20)23-12-10-19(21,5-2)6-3;;/h8-9,13H,4-7,10-12,18H2,1-3H3;2*1H |
| SMILES | Cl.Cl.CCCCOc1cc(ccc1N)C(=O)OCC[N+]([O-])(CC)CC |
| IUPAC Name | 2-(4-amino-3-butoxybenzoyl)oxy-N,N-diethylethanamine oxide;dihydrochloride |
| Show More |