Learn More
Oxolinic Acid Isopropyl Ester, Mikromol™
Product Code. 30051766
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051766 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30051766 Supplier Mikromol Supplier No. MM0898.050025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Oxolinic Acid Isopropyl Ester
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Oxolinic Acid Isopropyl Ester
CAS 2731414-28-9
Analyte or Component Names Oxolinic Acid Isopropyl Ester
Shipping Condition Room Temperature
Molecular Formula C16 H17 N O5
Certifications/Compliance ISO 9001
Synonym Oxolinic Acid Isopropyl Ester
InChI Formula InChI=1S/C16H17NO5/c1-4-17-7-11(16(19)22-9(2)3)15(18)10-5-13-14(6-12(10)17)21-8-20-13/h5-7,9H,4,8H2,1-3H3
SMILES CCN1C=C(C(=O)OC(C)C)C(=O)c2cc3OCOc3cc12
IUPAC Name propan-2-yl 5-ethyl-8-oxo-[1,3]dioxolo[4,5-g]quinoline-7-carboxylate
Molecular Weight (g/mol) 303.31
Quantity 25 mg
Formula Weight 303.1107 g/mol