Learn More
Oxcarbazepine EP Impurity F, Mikromol™
Product Code. 30052358
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052358 25 mg 25mg
30051981 100 mg 100mg
30461276 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30052358 Supplier Mikromol Supplier No. MM0637.100025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Oxcarbazepine EP Impurity F
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 10-Methoxy-5H-dibenzo[b,f]azepine-5-carbonyl Chloride
CAS 28721-08-6
Analyte or Component Names 10-Methoxy-5H-dibenzo[b,f]azepine-5-carbonyl Chloride
Shipping Condition Room Temperature
Molecular Formula C16 H12 Cl N O2
Certifications/Compliance ISO 17025
Synonym 10-Methoxy-5H-dibenz[b,f]azepine-5-carbonyl chloride
InChI Formula InChI=1S/C16H12ClNO2/c1-20-15-10-11-6-2-4-8-13(11)18(16(17)19)14-9-5-3-7-12(14)15/h2-10H,1H3
SMILES COC1=Cc2ccccc2N(C(=O)Cl)c3ccccc13
IUPAC Name 5-methoxybenzo[b][1]benzazepine-11-carbonyl chloride
Molecular Weight (g/mol) 286
Quantity 25 mg