Learn More
Oxatomide N-Oxide, Mikromol™
Description
Oxatomide N-Oxide
Specifications
Specifications
| Chemical Name or Material | Oxatomide N-Oxide |
| CAS | 2726534-33-2 |
| Analyte or Component Names | Oxatomide N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C27 H30 N4 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Oxatomide N-Oxide |
| InChI Formula | InChI=1S/C27H30N4O2/c32-27-28-24-14-7-8-15-25(24)30(27)16-9-19-31(33)20-17-29(18-21-31)26(22-10-3-1-4-11-22)23-12-5-2-6-13-23/h1-8,10-15,26H,9,16-21H2,(H,28,32) |
| SMILES | [O-][N+]1(CCCN2C(=O)Nc3ccccc23)CCN(CC1)C(c4ccccc4)c5ccccc5 |
| IUPAC Name | 3-[3-(4-benzhydryl-1-oxidopiperazin-1-ium-1-yl)propyl]-1H-benzimidazol-2-one |
| Show More |