missing translation for 'onlineSavingsMsg'
Learn More

Oxamflatin, MedChemExpress

Product Code. p-200100378
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16413698 10 mg 10mg
16406437 5 mg 5mg
16402836 25 mg 25mg
16400979 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16413698 Supplier Medchem Express Supplier No. HY102033

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oxamflatin (Metacept-3) is a potent HDAC inhibitor with an IC50 of 15.7 nM.

Oxamflatin (Metacept-3) is a potent HDAC inhibitor with an IC50 of 15.7 nM.

Specifications

Chemical Name or Material Oxamflatin
CAS 151720-43-3
Color Deep Yellow
Purity Grade Notes Research
Molecular Formula C17H14N2O4S
Quantity 10 mg
Synonym Metacept-3
Solubility Information DMSO : 50 mg/mL (146.04 mM; Need ultrasonic)
SMILES O=C(NO)/C=C/C#CC1=CC=CC(NS(=O)(C2=CC=CC=C2)=O)=C1
Molecular Weight (g/mol) 342.37
Formula Weight 342.37
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.