missing translation for 'onlineSavingsMsg'
Learn More

Oxamflatin, MedChemExpress

Product Code. 16402836
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16402836

Brand: Medchem Express HY102033

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oxamflatin (Metacept-3) is a potent HDAC inhibitor with an IC50 of 15.7 nM.

Oxamflatin (Metacept-3) is a potent HDAC inhibitor with an IC50 of 15.7 nM.

Specifications

Chemical Name or Material Oxamflatin
CAS 151720-43-3
Color Deep Yellow
Purity Grade Notes Research
Molecular Formula C17H14N2O4S
Quantity 25 mg
Synonym Metacept-3
Solubility Information DMSO : 50 mg/mL (146.04 mM; Need ultrasonic)
SMILES O=C(NO)/C=C/C#CC1=CC=CC(NS(=O)(C2=CC=CC=C2)=O)=C1
Molecular Weight (g/mol) 342.37
Formula Weight 342.37
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.