Learn More
Organophosphorous Pesticides Mixture 260 100 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Dichlorvos∣Dimethoate∣Malathion∣Parathion-ethyl∣Parathion-methyl |
| CAS | 62-73-7, 60-51-5, 121-75-5, 56-38-2, 298-00-0 |
| Analyte or Component Names | Dichlorvos∣Dimethoate∣Malathion∣Parathion-ethyl∣Parathion-methyl |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Dichlorvos 100 μg/mL∣Dimethoate 100 μg/mL∣Malathion 100 μg/mL∣Parathion-ethyl 100 μg/mL∣Parathion-methyl 100 μg/mL |