Learn More
Opipramol N-Oxide, Mikromol™
Description
Opipramol N-Oxide
Specifications
Specifications
| Chemical Name or Material | Opipramol N-Oxide |
| CAS | 2732277-08-4 |
| Analyte or Component Names | Opipramol N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H29 N3 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Opipramol N-Oxide |
| InChI Formula | InChI=1S/C23H29N3O2/c27-19-18-26(28)16-14-24(15-17-26)12-5-13-25-22-8-3-1-6-20(22)10-11-21-7-2-4-9-23(21)25/h1-4,6-11,27H,5,12-19H2 |
| SMILES | OCC[N+]1([O-])CCN(CCCN2c3ccccc3C=Cc4ccccc24)CC1 |
| IUPAC Name | 2-[4-(3-benzo[b][1]benzazepin-11-ylpropyl)-1-oxidopiperazin-1-ium-1-yl]ethanol |
| Show More |