Learn More
Olanzapine N-Oxide, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Olanzapine N-Oxide |
| CAS | 174794-02-6 |
| Analyte or Component Names | Olanzapine N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H20 N4 O S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Olanzapine Imp. D (EP), Olanzapine USP Related Compound C, Olanzapine USP RC C, 1-Methyl-4-(2-methyl-10H-thieno[2,3-b][1,5]benzodiazepin-4-yl)piperazin-1-oxide, Olanzapine N-Oxide, 2-Methyl-4-(4-methyl-4-oxidopiperazin-1-yl)-10H-thieno[2,3-b][1,5]-benzodiazepine, 2-Methyl-4-(4-methyl-4-oxido-1-piperazinyl)-10H-thieno[2,3-b][1,5]benzodiazepine, LY 170238, Olanzapine Related Compound C, Olanzapine Impurity D |
| InChI Formula | InChI=1S/C17H20N4OS/c1-12-11-13-16(20-7-9-21(2,22)10-8-20)18-14-5-3-4-6-15(14)19-17(13)23-12/h3-6,11,19H,7-10H2,1-2H3 |
| Solution Type | Neat |
| SMILES | Cc1cc2C(=Nc3ccccc3Nc2s1)N4CC[N+](C)([O-])CC4 |
| Show More |