Learn More
Octocrylene, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Octocrylene |
| CAS | 6197-30-4 |
| Analyte or Component Names | Octocrylene |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H27 N O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-Cyano-3,3-diphenyl-2-propenoic acid 2-ethylhexyl ester, 2-Ethylhexyl 2-cyano-3,3-diphenyl-prop-2-enoate, Octocrylene, 2-Ethylhexyl 2-cyano-3,3-diphenylacrylate, Parsol 340 |
| InChI Formula | InChI=1S/C24H27NO2/c1-3-5-12-19(4-2)18-27-24(26)22(17-25)23(20-13-8-6-9-14-20)21-15-10-7-11-16-21/h6-11,13-16,19H,3-5,12,18H2,1-2H3 |
| Solution Type | Neat |
| SMILES | CCCCC(CC)COC(=O)C(=C(c1ccccc1)c2ccccc2)C#N |
| Show More |