Learn More
Cytiva O-ring
Product Code. p-8004336 Shop All Cytiva Products
Click to view available options
Size:
11.3 x 2.4mm
27.1 x 1.60 mm
37.47 x 5.34mm
6.75 x 1.78mm
Unit Size:
Each
Pack of 10
Pack of 5
Product Code. 11785185
Brand: Cytiva 18110497
Only null left
Add to basket
Add to basket
Use in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules. Cytiva Life Sciences™ O-rings are made of nitrile rubber.
  • Can be used in aqueous solutions
  • Nearly all organic solvents
  • The following few exceptions: chlorinated hydrocarbons, acetone and otherketones, aliphatic esters and phenol. Solutions containingmore than 10% NaOH, 10% HCl, other strong mineral acidsor 5% acetic acid should not be used
  • Made of Nitrile Rubber (NBR)
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2/Pk.
Product Type O-Ring
Material Perfluor-Rubber
Size 11.3 x 2.4mm