Learn More
Cytiva O-ring
Shop All Cytiva Products
Click to view available options
Size:
11.3 x 2.4mm
27.1 x 1.60 mm
37.47 x 5.34mm
6.75 x 1.78mm
Unit Size:
Each
Pack of 10
Pack of 5
Description
- Can be used in aqueous solutions
- Nearly all organic solvents
- The following few exceptions: chlorinated hydrocarbons, acetone and otherketones, aliphatic esters and phenol. Solutions containingmore than 10% NaOH, 10% HCl, other strong mineral acidsor 5% acetic acid should not be used
- Made of Nitrile Rubber (NBR)
Specifications
Specifications
| Quantity | 5/Pk. |
| Product Type | O-Ring |
| Material | Nitrile Rubber |
| Size | 27.1 x 1.60 mm |