Learn More
Tocris Bioscience™ NVS-CECR2-1, >98%
GREENER_CHOICE
Product Code. 30000760
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000760 10 mg 10mg
30000973 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30000760 Supplier Tocris Bioscience™ Supplier No. 5744/10
Please to purchase this item. Need a web account? Register with us today!
Potent and selective inhibitor of cat eye syndrome chromosome region, candidate 2 (CECR2)
Potent and selective CECR2 (cat eye syndrome chromosome region, candidate 2) inhibitor (IC50 = 47 nM). Selective for CECR2 over a panel of bromodomains.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1992047-61-6
Molecular Formula C27H37N5O2S
Specifications
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Formula Weight 495.68
Purity >98%
Molecular Weight (g/mol) 495.686
Chemical Name or Material N-Cyclopropyl-2-(propylsulfonyl)-6-(1-(2,2,6,6-tetramethylpiperidin-4-yl)-1H-indol-5-yl)pyrimidin-4-