accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SOONNHNNHNCH3CH3H3CH3C

CAS RN 1992047-61-6

IUPAC Name: N-cyclopropyl-2-(propane-1-sulfonyl)-6-[1-(2,2,6,6-tetramethylpiperidin-4-yl)-1H-indol-5-yl]pyrimidin-4-amine
Synonyms: N-cyclopropyl-2-(propane-1-sulfonyl)-6-[1-(2,2,6,6-tetramethylpiperidin-4-yl)indol-5-yl]pyrimidin-4-amine
Molecular Weight (g/mol): 495.69
Molecular Formula: C27H37N5O2S
InChi Key: XVECNLUKQDKOST-UHFFFAOYSA-N
SMILES: CCCS(=O)(=O)C1=NC(=CC(NC2CC2)=N1)C1=CC=C2N(C=CC2=C1)C1CC(C)(C)NC(C)(C)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results