Learn More
Cytiva Nucleus Membrane
Made from polycarbonate film, for precision analysis of track-etched manufacturing
Brand: Cytiva 10418318
Additional Details : Weight : 0.05000kg
Description
Made from polycarbonate film, for precision analysis of track-etched manufacturing
- Membrane with low protein adsorption
- Manufactured to have a uniform hole diameter
- Reduces the risk of contamination due to almost no elution
- High chemical resistance and resistance to high temperatures
- Low ash content and tare weight for quantitative analysis
- Smooth surface for easy observation of retained particles
Specifications
Polycarbonate | |
Air Analysis, Bioassay, Cell Biology, Environmental Analysis, Epi-Fluorescence Microscopy, EPA Test, Fuel Test, Parasitology, and Underwater Microbiology | |
Nucleus Membrane |
3 μm | |
25 Sheets | |
90 mm |