Learn More
Macherey-Nagel™ Nucleosil™ 100-5 C18 HD CC Guard Columns Cartridges
Product Code. 10286891 Shop All Macherey-Nagel Products
Change view
Click to view available options
Diameter (Metric) Inner:
4 mm
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
10286891 4 mm Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10286891 Supplier Macherey-Nagel™ Supplier No. 721853.40
Only null left
Add to basket
Add to basket
Specifications
Quantity 3 Pack
Particle Size 5 μm
Length (Metric) 8 mm
Pore Size 100Å
Type CC Guard Columns Cartridges
Diameter (Metric) Inner 4 mm
For Use With (Equipment) EC columns of 4 and 4.6 mm ID with guard column adapter EC (REF 721359)
Stationary Phase NUCLEOSIL™ 100-5 C18 HD