Learn More
Macherey-Nagel™ Nucleodur™ C18 Pyramid HPLC CC Guard Columns
Product Code. 12488587 Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric) Inner:
3 mm
4 mm
Particle Size:
3 μm
5 μm
Unit Size:
Each
Pack of 3
Product Code. 12488587
Brand: Macherey-Nagel™ 761854.30
Only null left
Add to basket
Add to basket
Specifications
Carbon Load 14%
pH 1 to 9
Particle Size 3 μm
Length (Metric) 8 mm
Pore Size 110Å
Type CC Guard Column
Diameter (Metric) Inner 3 mm
Stationary Phase NUCLEODUR™ C18 Pyramid
Mobile Phase Compatibility Acetonitrile/Water
Solvents Acetonitrile/Water
Packing Material Silica
Crosslinking Octadecyl
Column Format Guard Column
For Use With (Application) LC/MS due to low bleeding characteristics
Technique HPLC
Particle Shape Spherical
Hydrophobicity Yes
Endcapped Yes
USP Type L1