Learn More
Macherey-Nagel™ Nucleodur™ C18 Isis HPLC CC Guard Columns
Product Code. 12798922 Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric) Inner:
3 mm
4 mm
Particle Size:
3 μm
5 μm
Unit Size:
Pack of 3
Product Code. 12798922
Brand: Macherey-Nagel™ 761310.30
Please to purchase this item. Need a web account? Register with us today!
Specifications
Carbon Load 20%
Particle Size 5 μm
Length (Metric) 8 mm
Pore Size 110Å
Type CC Guard Column
Diameter (Metric) Inner 3 mm
Stationary Phase NUCLEODUR™ C18 Isis
Mobile Phase Compatibility Acetonitrile/Water
Solvents Acetonitrile/Water
Packing Material Silica
Crosslinking Octadecyl
Column Format Guard Column
For Use With (Application) LC/MS and HPLC at pH extremes
Technique HPLC
Particle Shape Spherical
Hydrophobicity No
Endcapped Yes
USP Type L1