Learn More
NSFL1C Mouse, anti-Human, CoraLite Plus 555, Clone: 1F12E12, Proteintech
Mouse Monoclonal Antibody
140.00 € - 450.00 €
Specifications
| Antigen | NSFL1C |
|---|---|
| Clone | 1F12E12 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 555 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18076855
|
Proteintech
CL555-68182-100UL |
100 μL |
450.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18038415
|
Proteintech
CL555-68182-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NSFL1C antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NSFL1C. This antibody recognizes human, mouse, rat, pig antigen. The NSFL1C antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 555.
Specifications
| NSFL1C | |
| 1000 μg/mL | |
| CoraLite Plus 555 | |
| Mouse | |
| 55968 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1F12E12 | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat, Pig | |
| dJ776F14.1, NSFL1 (p97) cofactor (p47), NSFL1 cofactor p47, NSFL1C, p47, p97 cofactor p47, UBX1, UBXD10, UBXN2C | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.