Learn More
NSFL1C Mouse, anti-Human, CoraLite Plus 555, Clone: 1F12E12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL555-68182-20UL
 View more versions of this product
Product Code. 18038415
  • 145.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
The NSFL1C antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NSFL1C. This antibody recognizes human, mouse, rat, pig antigen. The NSFL1C antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 555.
Specifications
Specifications
NSFL1C
1F12E12
CoraLite Plus 555
Mouse
Primary
Human, Mouse, Rat, Pig
155 kDa
IgG1
Immunofluorescence
1000 μg/mL
dJ776F14.1, NSFL1 (p97) cofactor (p47), NSFL1 cofactor p47, NSFL1C, p47, p97 cofactor p47, UBX1, UBXD10, UBXN2C
20 μL
55968
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.