Learn More
NSC117079, MedChemExpress
Product Code. 16467928
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16467928
Brand: Medchem Express HY19819
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress NSC117079 is a novel PHLPP inhibitor.
NSC117079 is a novel PHLPP inhibitor.
Specifications
Chemical Name or Material NSC117079
CAS 500363-63-3
Color Purple
Purity Grade Notes Research
Molecular Formula C20H15N3O7S2
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (105.60 mM; Need ultrasonic)
SMILES O=S(C(C(N)=C1C2=O)=CC(NC3=CC=CC(S(=O)(N)=O)=C3)=C1C(C4=C2C=CC=C4)=O)(O)=O
Molecular Weight (g/mol) 473.48
Formula Weight 473.48
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation