missing translation for 'onlineSavingsMsg'
Learn More

NOP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen NOP2
Concentration 0.13 mg/mL
Applications Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16846843
View Documents
Proteintech
10448-1-AP-20UL
20 μL
145.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16836843
View Documents
Proteintech
10448-1-AP-150UL
150 μL
410.00 €
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

May play a role in the regulation of the cell cycle and the increased nucleolar activity that is associated with the cell proliferation. May act as ribosomal RNA methyltransferase.
Specifications

Specifications

NOP2
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Rat, Mouse
P46087, Q922K7
110109, 314969, 4839
NOP2 Fusion Protein Ag0498
Primary
-20°C
NOP2
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
NOL1, NOP120, NOP2, NSUN1, Nucleolar protein 1, Nucleolar protein 2 homolog, p120
Nop2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.