missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10448-1-AP-150UL
This item is not returnable.
View return policy
Description
May play a role in the regulation of the cell cycle and the increased nucleolar activity that is associated with the cell proliferation. May act as ribosomal RNA methyltransferase.Specifications
| NOP2 | |
| Polyclonal | |
| Unconjugated | |
| NOP2 | |
| NOL1, NOP120, NOP2, NSUN1, Nucleolar protein 1, Nucleolar protein 2 homolog, p120 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 110109, 314969, 4839 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P46087, Q922K7 | |
| Nop2 | |
| NOP2 Fusion Protein Ag0498 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction