Learn More
Amarell™ Nonmercury Enclosed Scale Low Temperature Precision Thermometer
Precision thermometer for the measurement of extremely cold temperatures.
Brand: Amarell™ L33012
Additional Details : Weight : 0.00100kg
Description
- Enclosed milk glass scale
- Toluene or pentane compound filling with a clear or blue capillary back
- Richter-top finish with button
- In transparent plastic square case
- Suitable for government verification or calibration
Specifications
N/A | |
300 mm | |
Toluene, Clear Capillary Back |
Total Immersion | |
Glass | |
Low Temperature Thermometer |