Learn More
Amarell™ Nonmercury Enclosed Scale Low Temperature Precision Thermometer
Precision thermometer for the measurement of extremely cold temperatures.
103.00 € - 117.00 €
Specifications
Immersion Type Total Immersion
Material Glass
Product Type Low Temperature Thermometer
View More Specs
Products 2
Product Code Brand Thermometer Fill Length (Metric) Price Quantity & Availability  
Product Code Brand Thermometer Fill Length (Metric) Price Quantity & Availability  
10717981
View Documents Promotion Details
Amarell™
L33018
Pentane Compound, Blue Capillary Back 350 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10012933
View Documents Promotion Details
Amarell™
L33012
Toluene, Clear Capillary Back 300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Enclosed milk glass scale
  • Toluene or pentane compound filling with a clear or blue capillary back
  • Richter-top finish with button
  • In transparent plastic square case
  • Suitable for government verification or calibration
Specifications
Specifications
Total Immersion
Glass
Low Temperature Thermometer
Videos
SDS
Documents
Documents
Product Certifications