Learn More
Hofmann Klaus™ Nitrogen Schlenk Tube
Brand:  Hofmann Klaus™ 20510.50
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10186580
  • 67.95 € / Each
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
Description
Description
Specifications
Specifications
Nitrogen Tube with Side Stopcock, with Solid Glass Plug and Hose Connection
50 mL
Neck with Standard Ground Socket (14/23)