Learn More
Hofmann Klaus™ Nitrogen Schlenk Tube
Brand: Hofmann Klaus™ 20510.50
Additional Details : Weight : 0.25000kg
Specifications
Nitrogen Tube with Side Stopcock, with Solid Glass Plug and Hose Connection | |
50 mL | |
Neck with Standard Ground Socket (14/23) |