Learn More
Hofmann Klaus™ Nitrogen Schlenk Tube
63.18 € - 73.78 €
Product Code | Brand | Description | Type | Capacity (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Type | Capacity (Metric) | Price | Quantity & Availability | |||
10430991
|
Hofmann Klaus™
20511.29 |
Nitrogen Tube with Side Stopcock, with Solid Glass Plug and Hose Connection | Neck with Standard Ground Cone (14/23) | 10 mL |
64.34 €
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||
10547302
|
Hofmann Klaus™
20513.61 |
Nitrogen Tube with Valve Stopcock | Neck with Standard Ground Cone (14/23) | 100 mL |
73.78 €
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||
10379700
|
Hofmann Klaus™
20510.29 |
Nitrogen Tube with Side Stopcock, with Solid Glass Plug and Hose Connection | Neck with Standard Ground Socket (14/23) | 10 mL |
64.24 €
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||
10389700
|
Hofmann Klaus™
20510.38 |
Nitrogen Tube with Side Stopcock, with Solid Glass Plug and Hose Connection | Neck with Standard Ground Socket (14/23) | 25 mL |
71.97 €
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||
10186580
|
Hofmann Klaus™
20510.50 |
Nitrogen Tube with Side Stopcock, with Solid Glass Plug and Hose Connection | Neck with Standard Ground Socket (14/23) | 50 mL |
67.95 €
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||
10596912
|
Hofmann Klaus™
20513.38 |
Nitrogen Tube with Valve Stopcock | Neck with Standard Ground Cone (14/23) | 25 mL |
63.18 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||