Learn More
Niflumic Acid Isopropyl Ester, Mikromol™
Product Code. 30052612
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052612 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30052612 Supplier Mikromol Supplier No. MM0850.020025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Niflumic Acid Isopropyl Ester
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Niflumic Acid Isopropyl Ester
CAS 2732347-70-3
Analyte or Component Names Niflumic Acid Isopropyl Ester
Shipping Condition Room Temperature
Molecular Formula C16 H15 F3 N2 O2
Certifications/Compliance ISO 9001
Synonym Niflumic Acid Isopropyl Ester
InChI Formula InChI=1S/C16H15F3N2O2/c1-10(2)23-15(22)13-7-4-8-20-14(13)21-12-6-3-5-11(9-12)16(17,18)19/h3-10H,1-2H3,(H,20,21)
SMILES CC(C)OC(=O)c1cccnc1Nc2cccc(c2)C(F)(F)F
IUPAC Name propan-2-yl 2-[3-(trifluoromethyl)anilino]pyridine-3-carboxylate
Molecular Weight (g/mol) 324.3
Quantity 25 mg
Formula Weight 324.1086 g/mol