missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nestin Antibody (4D11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 17 publications
213.00 € - 588.00 €
Specifications
| Antigen | Nestin |
|---|---|
| Clone | 4D11 |
| Concentration | 1 mg/ml |
| Dilution | Western Blot 1:5000, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18087956
|
Novus Biologicals
NBP1-92717SS |
0.025 mL |
213.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18786573
|
Novus Biologicals
NBP1-92717 |
0.1 mL |
588.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nestin Monoclonal specifically detects Nestin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Nestin | |
| 1 mg/ml | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers | |
| 50% PBS, 50% glycerol with 5mM Sodium Azide | |
| FLJ21841, nestin | |
| NES | |
| IgG1 | |
| Protein G purified | |
| 240 kDa |
| 4D11 | |
| Western Blot 1:5000, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Rat | |
| P48681 | |
| 10763 | |
| Recombinant construct, amino acids 317-630 of the human Nestin protein expressed in and purified from E. coli. [UniProt P48681] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title