missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nestin Antibody (4D11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 17 publications
Brand: Novus Biologicals NBP1-92717
This item is not returnable.
View return policy
Description
Nestin Monoclonal specifically detects Nestin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Nestin | |
| Monoclonal | |
| 1 mg/ml | |
| Western Blot 1:5000, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500 | |
| P48681 | |
| NES | |
| Recombinant construct, amino acids 317-630 of the human Nestin protein expressed in and purified from E. coli. [UniProt P48681] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Ascites |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 4D11 | |
| Unconjugated | |
| 50% PBS, 50% glycerol with 5mM Sodium Azide | |
| FLJ21841, nestin | |
| Mouse | |
| 240 kDa | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers | |
| 10763 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction