missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NBR1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | NBR1 |
|---|---|
| Concentration | 0.42 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Flow Cytometry, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16815444
|
Proteintech
16004-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16805444
|
Proteintech
16004-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene was originally identified as an ovarian tumor antigen monitored in ovarian cancer. The encoded protein contains a B-box/coiled coil motif, which is present in many genes with transformation potential, but the function of this protein is unknown. This gene is located on a region of chromosome 17q21.1 that is in close proximity to tumor suppressor gene BRCA1. Three alternatively spliced variants encoding the same protein have been identified for this gene.Specifications
| NBR1 | |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Non-human Primate, Mouse | |
| P97432, Q14596, Q501R9 | |
| 17966, 303554, 4077 | |
| NBR1 Fusion Protein Ag8652 | |
| Primary | |
| -20°C | |
| NBR1 |
| 0.42 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 1A1 3B, 1A13B, KIAA0049, M17S2, MIG19, NBR1, neighbor of BRCA1 gene 1, Next to BRCA1 gene 1 protein, Protein 1A1 3B | |
| Nbr1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title