missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NBR1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16004-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene was originally identified as an ovarian tumor antigen monitored in ovarian cancer. The encoded protein contains a B-box/coiled coil motif, which is present in many genes with transformation potential, but the function of this protein is unknown. This gene is located on a region of chromosome 17q21.1 that is in close proximity to tumor suppressor gene BRCA1. Three alternatively spliced variants encoding the same protein have been identified for this gene.Specifications
| NBR1 | |
| Polyclonal | |
| Unconjugated | |
| NBR1 | |
| 1A1 3B, 1A13B, KIAA0049, M17S2, MIG19, NBR1, neighbor of BRCA1 gene 1, Next to BRCA1 gene 1 protein, Protein 1A1 3B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17966, 303554, 4077 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Flow Cytometry, Immunoprecipitation | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P97432, Q14596, Q501R9 | |
| Nbr1 | |
| NBR1 Fusion Protein Ag8652 | |
| 20 μL | |
| Primary | |
| Rat, Human, Non-human Primate, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction