Learn More
N3-(6-Methoxyquinolin-8-yl)pentane-1,3-diamine (Secaquine), TRC
Product Code. 30483540
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483540 100 mg 100mg
30471260 25 mg 25mg
30481336 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30483540 Supplier Toronto Research Chemicals Supplier No. TRCM910003100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2732404-25-8
Molecular Formula C15 H21 N3 O
Molecular Weight (g/mol) 259.347
Synonym Secaquine
IUPAC Name 3-N-(6-methoxyquinolin-8-yl)pentane-1,3-diamine
SMILES CCC(CCN)Nc1cc(OC)cc2cccnc12
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C15H21N3O/c1-3-12(6-7-16)18-14-10-13(19-2)9-11-5-4-8-17-15(11)14/h4-5,8-10,12,18H,3,6-7,16H2,1-2H3
Formula Weight 259.168
Chemical Name or Material N3-(6-Methoxyquinolin-8-Yl)Pentane-1,3-Diamine (Secaquine)