CAS RN 2732404-25-8
CAS RN 2732404-25-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N3-(6-Methoxyquinolin-8-yl)pentane-1,3-diamine (Secaquine), TRC
CAS: 2732404-25-8 Molecular Formula: C15 H21 N3 O Molecular Weight (g/mol): 259.347 Synonym: Secaquine IUPAC Name: 3-N-(6-methoxyquinolin-8-yl)pentane-1,3-diamine SMILES: CCC(CCN)Nc1cc(OC)cc2cccnc12