Learn More
N-[(R)-(4-Nitrophenoxy)phenoxyphosphinyl]-L-alanine 2-Ethylbutyl Ester, Mikromol™
Description
N-[(R)-(4-Nitrophenoxy)phenoxyphosphinyl]-L-alanine 2-Ethylbutyl Ester
Specifications
Specifications
| Chemical Name or Material | N-[(R)-(4-Nitrophenoxy)phenoxyphosphinyl]-L-alanine 2-Ethylbutyl Ester |
| CAS | 1354823-37-2 |
| Analyte or Component Names | N-[(R)-(4-Nitrophenoxy)phenoxyphosphinyl]-L-alanine 2-Ethylbutyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H27 N2 O7 P |
| Certifications/Compliance | ISO 9001 |
| Synonym | L-Alanine, N-[(R)-(4-nitrophenoxy)phenoxyphosphinyl]-, 2-ethylbutyl ester, N-[(R)-(4-Nitrophenoxy)phenoxyphosphinyl]-L-alanine 2-Ethylbutyl Ester |
| InChI Formula | InChI=1S/C21H27N2O7P/c1-4-17(5-2)15-28-21(24)16(3)22-31(27,29-19-9-7-6-8-10-19)30-20-13-11-18(12-14-20)23(25)26/h6-14,16-17H,4-5,15H2,1-3H3,(H,22,27)/t16-,31+/m0/s1 |
| SMILES | CCC(CC)COC(=O)[C@H](C)N[P@@](=O)(Oc1ccccc1)Oc2ccc(cc2)[N+](=O)[O-] |
| IUPAC Name | 2-ethylbutyl (2S)-2-[[(4-nitrophenoxy)-phenoxyphosphoryl]amino]propanoate |
| Show More |