Learn More
n-Propyl hexanoate, 98%, Thermo Scientific™
Product Code. 11420718
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420718 25 g 25g
11430718 100 g 100g
11440718 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11420718 Supplier Thermo Scientific Alfa Aesar Supplier No. B25215.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 626-77-7
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00053803
InChI Key HTUIWRWYYVBCFT-UHFFFAOYSA-N
Synonym propyl caproate, caproic acid propyl ester, hexanoic acid, propyl ester, n-propyl hexanoate, propionyl hexanoate, n-propyl n-hexanoate, hexanoic acid propyl ester, natural, fema no. 2949, n-propyl hexanoaten
PubChem CID 12293
ChEBI CHEBI:87365
IUPAC Name propyl hexanoate
SMILES CCCCCC(=O)OCCC
Specifications
Melting Point -69°C
Density 0.876
Boiling Point 187°C
Flash Point 52°C (126°F)
Odor Fruit-like
Linear Formula CH3(CH2)4CO2CH2CH2CH3
Refractive Index 1.412
Quantity 25 g
UN Number UN3272
Formula Weight 158.24
Percent Purity 98%
Chemical Name or Material n-Propyl hexanoate