CAS RN 626-77-7
CAS RN 626-77-7
IUPAC Name:
propyl hexanoate
Synonyms:
propyl hexanoate
Molecular Weight (g/mol):
158.24
Molecular Formula:
C9H18O2
InChi Key:
HTUIWRWYYVBCFT-UHFFFAOYSA-N
SMILES:
CCCCCC(=O)OCCC
1
–
1
of
1
results
1
n-Propyl hexanoate, 98%, Thermo Scientific™
CAS: 626-77-7 Molecular Formula: C9H18O2 Molecular Weight (g/mol): 158.24 MDL Number: MFCD00053803 InChI Key: HTUIWRWYYVBCFT-UHFFFAOYSA-N Synonym: propyl caproate,caproic acid propyl ester,hexanoic acid, propyl ester,n-propyl hexanoate,propionyl hexanoate,n-propyl n-hexanoate,hexanoic acid propyl ester,natural,fema no. 2949,n-propyl hexanoaten PubChem CID: 12293 ChEBI: CHEBI:87365 IUPAC Name: propyl hexanoate SMILES: CCCCCC(=O)OCCC