Learn More
N-Phenylacetyl-L-glutamine (Phenylacetylglutamine), Mikromol™
Description
N-Phenylacetyl-L-glutamine (Phenylacetylglutamine)
Specifications
Specifications
| Chemical Name or Material | Phenylacetyl L-Glutamine |
| CAS | 28047-15-6 |
| Analyte or Component Names | Phenylacetyl L-Glutamine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H16 N2 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | N2-(2-Phenylacetyl)-L-glutamine, N-Phenylacetylglutamine, N2-(Phenylacetyl)-L-glutamine, NSC 203800, Phenylacetyl-L-glutamine, Phenylacetylglutamine, N-Phenylacetyl-L-glutamine |
| InChI Formula | InChI=1S/C13H16N2O4/c14-11(16)7-6-10(13(18)19)15-12(17)8-9-4-2-1-3-5-9/h1-5,10H,6-8H2,(H2,14,16)(H,15,17)(H,18,19)/t10-/m0/s1 |
| SMILES | NC(=O)CC[C@H](NC(=O)Cc1ccccc1)C(=O)O |
| IUPAC Name | (2S)-5-amino-5-oxo-2-[(2-phenylacetyl)amino]pentanoic acid |
| Show More |