Learn More
N-Phenyl-N-[1-(2-phenylethyl)piperidin-4-yl]acetamide, Mikromol™
Description
N-Phenyl-N-[1-(2-phenylethyl)piperidin-4-yl]acetamide
Specifications
Specifications
| Chemical Name or Material | Acetyl Fentanyl |
| CAS | 3258-84-2 |
| Analyte or Component Names | Acetyl Fentanyl |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H26 N2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Fentanyl Imp. C (EP), Fentanyl USP Related Compound G CI, Fentanyl USP RC G CI, N-Phenyl-N-[1-(2-phenylethyl)piperidin-4-yl]acetamide, N-Phenyl-N-[1-(2-phenylethyl)-4-piperidinyl]acetamide, N-(1-Phenethyl-4-piperidyl)acetanilide, 1-Phenethyl-4-(N-phenylacetamido)piperidine, Acetylfentanyl, Fentanyl Related Compound G, Fentanyl Citrate Impurity C, Fentanyl Impurity C, Fentanyl Impurity C |
| InChI Formula | InChI=1S/C21H26N2O/c1-18(24)23(20-10-6-3-7-11-20)21-13-16-22(17-14-21)15-12-19-8-4-2-5-9-19/h2-11,21H,12-17H2,1H3 |
| SMILES | CC(=O)N(C1CCN(CCc2ccccc2)CC1)c3ccccc3 |
| IUPAC Name | N-phenyl-N-[1-(2-phenylethyl)piperidin-4-yl]acetamide |
| Show More |