Learn More
n-Octyl L-2-pyrrolidinone-5-carboxylate, 98%, Thermo Scientific™
Product Code. 11414887
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11414887
Brand: Thermo Scientific Alfa Aesar B21838.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4931-70-8
Molecular Formula C4H11N5
Molecular Weight (g/mol) 129.17
MDL Number MFCD00236053
InChI Key XZWYZXLIPXDOLR-UHFFFAOYSA-N
Synonym octyl 2s-5-oxopyrrolidine-2-carboxylate, s-octyl 5-oxopyrrolidine-2-carboxylate, octyl 5-oxo-l-prolinate, n-octyl l-2-pyrrolidinone-5-carboxylate, octyl pyroglutamate, 5-oxo-l-proline octyl ester, n-octyl l-2-pyrrolidone-5-carboxylate
PubChem CID 107610
IUPAC Name 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide
SMILES CN(C)C(=N)N=C(N)N
Specifications
Boiling Point 140°C (0.06 mmHg)
Refractive Index 1.472
Quantity 25 g
Beilstein 478315
Formula Weight 241.33
Percent Purity 98%
Chemical Name or Material n-Octyl L-2-pyrrolidinone-5-carboxylate