accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4931-70-8

CH3NCH3HNNH2NH2N

CAS RN 4931-70-8

IUPAC Name: 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide
Synonyms: metformin (inn) and its salts
Molecular Weight (g/mol): 129.17
Molecular Formula: C4H11N5
InChi Key: XZWYZXLIPXDOLR-UHFFFAOYSA-N
SMILES: CN(C)C(=N)N=C(N)N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results